Learn More
Description
- Reagents include Recovery Buffer and Loading Dye
- Loading Dye: 5X concentration; RNA native sample buffer
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
Specifications
| Concentration | 5X |
| Form | Liquid |
| Quantity | 5 x 1 mL |
| Storage Requirements | Room Temperature |
| Product Type | DNA Loading Dye |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.