Learn More
Lonza™ GelStar™ Photographic Filter
Description
- 3 in. square Wratten™ #9 filter optimizes detection sensitivity when using black and white photography on Polaroid™ camera-based systems
Specifications
Specifications
| For Use With (Equipment) | Gelstar Nucleic Acid Gel Stain |
| For Use With (Application) | The gelstar™ photographic filter is a 3 in. square wratten no. 9 filter designed to optimize detection sensitivity when using black and white photography on polaroid camera based systems |
| Product Type | Gelstar™ Gel Stain Photo Filter |