Learn More
Lonza™ IsoGel™ IEF Plate Accessories
Description
- Blotting paper is for use with precast agarose IEF plates
- Accessory pack includes masks, 100mm and 125mm wicks and blotting paper
- Accessory bulk pack includes 125mm wicks and blotting paper
Specifications
Specifications
| Separation Type | Isoelectric Focusing |
| Form | Dried |
| For Use With (Application) | Isoelectric focusing, Antibody separation and analysis |
| Storage Requirements | 18° to 26°C |
| Product Type | Electrophoresis Accessory Bulk Pack |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.