Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ ProSieve™ EX Running Buffer

Catalog No. BMA00200307 Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
1L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMA00200307 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMA00200307 Supplier Lonza™ Supplier No. 00200307
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Novel formulation of the classic Laemmli buffer

  • Proprietary solution to drastically increase the separation speed without excessive heat generation affecting the gel
  • Direct replacement for tris-glycine SDS running buffer
  • Compatible with commercial precast and hand cast gels

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Quantity 1L
Storage Requirements Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.