Learn More
Lonza™ Reliant™ Precast Gels
Description
- Manufactured with high-quality SeaKem™ Gold and NuSieve™ 3:1 Plus brand agaroses
- Easy to handle
- Fit most common mini submerged electrophoresis systems
- Available with 0.5μg/mL of ethidium bromide for ease of visualizing DNA bands during and after the run
Specifications
Specifications
| Buffer | TBE |
| Color | Clear |
| Concentration | 2% |
| Format | Precast Gel |
| Label or Dye | Ethidium Bromide |
| Separation Range | 100 to ≥2000 bp |
| Wells | 20-well |
| Dnase Free | Yes |
| Formulation | 2% SeaKem™ Gold Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide |
| For Use With (Application) | Rapid and reproducible resolution of DNA fragments from 8bp to 10kb |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.