Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ Reliant™ Precast Gels

Catalog No. BMA54944
Change view
Click to view available options
Separation Range:
100 to ≥2000 bp
100 to ≥3000 bp
20 to ≥1000 bp
300 to ≥8000 bp
400 to ≥10,000 bp
8 to ≥1000 bp
Buffer:
TAE
TBE
Wells:
12-well
20-well
24-well
8-well
Label or Dye:
Ethidium Bromide
None
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Separation Range Buffer Wells Label or Dye
BMA54944 8 to ≥1000 bp TBE 20-well None
BMA54801 400 to ≥10,000 bp TAE 8-well None
BMA54903 300 to ≥8000 bp TBE 8-well Ethidium Bromide
BMA54928 8 to ≥1000 bp TBE 20-well Ethidium Bromide
BMA54905 300 to ≥8000 bp TBE 24-well Ethidium Bromide
BMA54927 8 to ≥1000 bp TBE 8-well Ethidium Bromide
BMA54925 20 to ≥1000 bp TAE 8-well Ethidium Bromide
BMA54929 8 to ≥1000 bp TBE 24-well Ethidium Bromide
BMA54939 100 to ≥2000 bp TBE 20-well Ethidium Bromide
BMA54907 300 to ≥8000 bp TBE 20-well Ethidium Bromide
BMA54803 400 to ≥10,000 bp TAE 8-well Ethidium Bromide
BMA54820 300 to ≥8000 bp TBE 12-well Ethidium Bromide
BMA54813 100 to ≥3000 bp TBE 24-well Ethidium Bromide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Ethidium bromide is a powerful mutagen. Gloves should be worn when opening pouches and handling gel solutions containing ethidium bromide.
Catalog No. BMA54944 Supplier Lonza™ Supplier No. 54944
Add to Cart
Add to Cart
Ethidium bromide is a powerful mutagen. Gloves should be worn when opening pouches and handling gel solutions containing ethidium bromide.

For rapid and reproducible resolution of DNA sizes from 8bp-10kb - Lonza™ Reliant™ Precast Gels are ideal for analysis of PCR, restriction digest fragments or most cloning needs.

  • Manufactured with high-quality SeaKem™ Gold and NuSieve™ 3:1 Plus brand agaroses
  • Easy to handle
  • Fit most common mini submerged electrophoresis systems
  • Available with 0.5μg/mL of ethidium bromide for ease of visualizing DNA bands during and after the run
TRUSTED_SUSTAINABILITY

Specifications

Buffer TBE
Color Clear
Concentration 4%
Format Precast Gel
Label or Dye None
Separation Range 8 to ≥1000 bp
Wells 20-well
Dnase Free Yes
Formulation 4% NuSieve 3:1 Plus Agarose, TBE Buffer
For Use With (Application) Rapid and reproducible resolution of DNA fragments from 8bp to 10kb
Material Agarose
Quantity 20 Gels
RNase Free Yes
Storage Requirements 18° to 26°C
Thickness (English) 0.19 in.
Thickness (Metric) 0.5 cm
Volume Per Well ≤15 μL
Length (English) Gel 3.7 in.
Length (Metric) Gel 9.5 cm
Width (English) Gel 2.37 in.
Width (Metric) Gel 6 cm
Product Type Agarose Gel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.