Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ SimplyLoad™ DNA Ladders

Catalog No. BMA50327
Change view
Click to view available options
No. of Bands:
10
21
Concentration:
32 ng/μL
60 ng/μL
Increments:
100bp
100bp and 1000bp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Bands Concentration Increments
BMA50327 10 32 ng/μL 100bp
BMA50333 21 60 ng/μL 100bp and 1000bp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BMA50327 Supplier Lonza™ Supplier No. 50327
Add to Cart
Add to Cart

Ready-to-load markers contain loading buffer to save time and pipetting steps

•    Consist of RNA transcripts in sizes of 0.5, 1, 1.5, 2, 2.5, 3, 4, 5, 6, and 9kb
•    Can be denatured with standard procedures
•    Can be visualized on Northern blots with labeled lambda sequence
•    Can be detected with ethidium bromide or nucleic acid stains
•    Store at -20°C for up to six months or at -80°C for up to 24 months

TRUSTED_SUSTAINABILITY

Specifications

Concentration 32 ng/μL
Form Liquid
Formulation 50% GC content
For Use With (Application) Gel electrophoresis
Increments 100bp
No. of Bands 10
Quantity 500 μL
Storage Requirements 2° to 8°C
Product Type DNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.