Learn More
United Scientific Supplies Loop The Loop Track
SDPSCI_ED
Catalog No. S27826 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Loop The Loop Track
Catalog No.
S27826 Loop The Loop Track
1 options
Catalog No. S27826 Supplier United Scientific Supplies Supplier No. LOOP01
Add to Cart
Add to Cart
Demonstrates centripetal acceleration and the transformation of potential energy to kinetic energy
Specifications
Product Type Loop The Loop Track
For Use With (Application) Motion Experiment
Includes Sturdy Base with Rubber Feet, Metal Ramp with Loop, 16/19 mm Steel Balls, Activity Guide