Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Loratadine, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18603972 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-972 25 g Poly Pail
18-603-970 1 g Poly Pail
18-603-973 100 g Poly Pail
18-603-968 1 kg Poly Pail
18-603-969 5 kg Poly Pail
18-603-971 5 g Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-603-972 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L301025GM

L3010, 79794-75-5, C22H23N2O2Cl

Spectrum™ Chemical Loratadine, USP is an antihistamine that treats the typical symptoms of allergies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 79794-75-5
Molecular Formula C22H23ClN2O2
Molecular Weight (g/mol) 382.89
InChI Key JCCNYMKQOSZNPW-UHFFFAOYSA-N
IUPAC Name ethyl 4-{13-chloro-4-azatricyclo[9.4.0.03,8]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}piperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(CC1)=C1C2=CC=C(Cl)C=C2CCC2=CC=CN=C12

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Poly Pail
Quantity 25 g
Percent Purity 98.5 to 101%
Grade USP
Physical Form Powder or Waxy Solid
Chemical Name or Material Loratadine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.