Learn More
Enzo Life Sciences Lorcainide. HCl (50mg). CAS: 58934-46-6
SDP
Supplier:  Enzo Life Sciences BMLNA1430050
Encompass_Preferred
Possesses both local anesthetic and antiarrhythmic properties (class 1C agent). Elevates the ventricular fibrillation threshold and protects against ouabain-induced arrhythmias in animal models. Its mechamism of action is primarly inhibition of sodium channels in a voltage- and frequency-dependent fashion. Also inhibits the microsomal Na+,K+-ATPase in a concentration-dependent manner (IC50=33.5µM) . Reduces neuronal death in cellular models of hypoxic injury when combined with a glutamate receptor antagonist and blocks aconitine-induced mortality in a rat model (ED50=30mg/kg). Alternative name: N-(4-Chlorophenyl)-N-[1-(1-isopropyl)-4-piperidinyl]phenylacetamide . HCl. Purity: ≥98% (TLC). Solubility: Soluble in water (>30mg/ml), DMSO (>25mg/ml) or 100% ethanol (>25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1188
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.