Learn More
Apexbio Technology LLC Phenylephrine HCl 61-76-7 100mg
SDP
Supplier:  Apexbio Technology LLC B1344100
Encompass_Preferred
Phenylephrine HCl (CAS 61-76-7) is a selective agonist of 1-adrenergic receptors a class of G protein-coupled receptors that mediate the effects of endogenous catecholamines such as norepinephrine and epinephrine Comparative studies indicate phenylephrine displays significant subtype selectivity with inhibitory pKi values of 4 87 4 70 and 5 86 for 1B- 1C- and 1A/D-adrenergic receptors respectively and agonist potencies (pEC50) of 6 47 6 77 and 7 01 for these subtypes In preclinical models phenylephrine has been shown to enhance alveolar fluid clearance and promote pulmonary edema absorption in ventilator-induced lung injury in a dose-dependent manner This compound is widely utilized in studies of adrenergic receptor signaling and pulmonary physiology
Catalog No. 50-000-92516
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.