Learn More
Selleck Chemical LLC Losmapimod (GW856553X) 10mg 585543-15-3 GW856553, GSK-AHAB
SDP
Supplier:  Selleck Chemical LLC S721510MG
Encompass_Preferred
Losmapimod (GW856553X, GW856553, GSK-AHAB) is a selective, potent, and orally active p38 MAPK inhibitor with pKi of 8.1 and 7.6 for p38? and p38?, respectively. P38 MAPKs are involved in cell differentiation, apoptosis and autophagy. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4528
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.