Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Lot Certified Graduated Microcentrifuge Tube

Catalog No. 22010089
Change view
Click to view available options
Volume (Metric):
1.5 mL
2.0 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-010-089 1.5 mL
22-010-092 2.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-010-089 Supplier Globe Scientific Supplier No. 111562
Add to Cart
Add to Cart

RNase Free, DNase Free, Human DNA Free, Pyrogen Free, ATP Free and PCR Inhibitor Free

  • RNase free, DNase free, human DNA free, Pyrogen free, ATP free and PCR inhibitor free
  • Made of polypropylene (PP)
  • With attached leak-resistant snap cap
  • Molded with precise graduation marks for accurate measurements

Specifications

Autoclavable Autoclavable
Color Natural
Material Polypropylene
Increments 0.5 mL
No. per Pack 500
Volume (Metric) 1.5 mL
Graduated Yes
Closure Type Snap Cap
Max. RCF 20000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.