Learn More
Medchemexpress LLC HY-N1505 5mg Medchemexpress, Loureirin A CAS:119425-89-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN15055MG
Encompass_Preferred
Medchemexpress, HY-N1505 5mg Loureirin A CAS:119425-89-7 Loureirin A is a flavonoid extracted from Dragon's Blood, can inhibit Akt phosphorylation, and has antiplatelet activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3849