Learn More
Selleck Chemical LLC Lovastatin 50mg 75330-75-5 MK-803
SDP
Supplier:  Selleck Chemical LLC S206150MG
Encompass_Preferred
Lovastatin (MK-803) is an inhibitor of HMG-CoA reductase with IC50 of 3.4 nM in a cell-free assay, used for lowering cholesterol (hypolipidemic agent). Lovastatin triggers autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1370080
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.