Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for total alkalinity using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
| For Use With (Application) | Testing water with the Aqua comparator™ |
| For Use With (Equipment) | Aqua comparator™ |
| Range | 20 to 240mg/L |
| Type | Total Alkalinity Color Disc |