Learn More
Lovibond™ Color of Water Comparator Disc for 700 Nesslerizer Comparator
Shop All Tintometer Group ProductsDescription
Color of Water Comparator Disc for 700 Nesslerizer Comparator
Specifications
Specifications
| For Use With (Application) | Measuring the color of potable water in terms of an easily reproducible color standard |
| For Use With (Equipment) | 700 Nesslerizer comparator |
| Range | 30 to 70 Color Units |
| Test Method | Pt-Co or APHA |
| Type | Color of Water Comparator Disc |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.