Learn More
Description
These rugged, waterproof, single parameter colorimeters are ideal for laboratory and field testing
- Ranges: 0-2.0mg/L and 0-8.0mg/L
- pH range: 6.5 to 8.5 pH units
- Method uses DPD reagent for chlorine and Phenol Red Solution for pH
- Free and total chlorine powder packs included (50 tests each)
- pH phenol red solution (15mL) included
- Large, backlit display; real time clock; data logging capabilities
- Ability to export data using the IRiM
Specifications
Specifications
| Accuracy | 3% Full scale |
| Calibration | Yes |
| Certifications/Compliance | CE |
| Data Storage | Automatic; 16 data sets with time and date stamp |
| Display | Backlit LCD |
| Dimensions (L x W x H) | 6.1 x 3 x 1.5 in. (15.5 x 7.6 x 3.8 cm) |
| IP Rating | IP68 |
| Light Source | LED |
| Type | Chlorine and pH Colorimeter |
| For Use With (Application) | Laboratory and field testing |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.