Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Liquid Reagents for Water Analysis

Catalog No. 09534457 Shop All Tintometer Group Products
Change view
Click to view available options
Color:
Colorless
Dark Orange
pH Range:
8
8.6
13.7
Volume (English):
1.01 oz.
16.9 oz.
Volume (Metric):
30 mL
500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color pH Range Volume (English) Volume (Metric)
09-534-457 Dark Orange 8 16.9 oz. 500 mL
09-534-455 Colorless 13.7 16.9 oz. 500 mL
09-534-458 Colorless 8.6 1.01 oz. 30 mL
09-534-459 Colorless 8.6 16.9 oz. 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09-534-457 Supplier Lovibond™ Supplier No. 530632
Add to Cart
Add to Cart

Specifications

Color Dark Orange
pH Range 8
Volume (Metric) 500 mL
Format Liquid
For Use With (Application) Water analysis
For Use With (Equipment) SC450 manganese, MC500 colorimeter, SP600 spectrophotometer, manganese reagent set
Range 0 to 0.7mg/L
Storage Requirements 20°C
Type PAN Indicator Solution
Volume (English) 16.9 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.