Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Model 711-A Nesslerizer System Nessler Tubes

Catalog No. 09534635 Shop All Tintometer Group Products
Change view
Click to view available options
Length (Metric):
250 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
09-534-635 250 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-635 Supplier Lovibond™ Supplier No. 354200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Set of two clear glass Nessler tubes

  • Water sample tubes
  • Path length: 250 mm (9.8 in.)
  • Plungers included
  • Use for water color comparison
  • Compatible with: Lovibond Nessslerizer System 711-A1 (FS Cat. No. 15-405-311)

Specifications

Material Glass
Color Clear
Length (Metric) 250 mm
For Use With (Equipment) 711-A1 Orbeco-Hellige™/Lovibond™ nessslerizer system
For Use With (Application) Water color comparison
Includes Plungers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.