Learn More
Description
- With graduation
- Material: Low density polyethylene
- Frapid transfer of liquids
- For single use only
Specifications
Specifications
| Material | LDPE |
| Sterility | Non-sterile |
| For Use With (Application) | Rapid transfer of liquids |
| Graduated | Yes |
| Graduations | 0.5 to 2.5 mL |
| Quantity | 1500/Cs. |
| Packaging Type | 500/bag |
| Volume (Metric) | 2.5 mL |
| Length (Metric) | 153 mm |
| Type | Serum Pipette |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
