Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tube
GREENER_CHOICE

Catalog No. 01182577 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
0.6 mL
1.5 mL
2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-182-577 0.6 mL
01-182-572 1.5 mL
01-182-576 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-182-577 Supplier Thermo Scientific™ Supplier No. 3403DLB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Microcentrifuge Tubes ensure optimal sample integrity with minimal DNA loss. These tubes provide secure closure and reliable performance, perfect for sensitive applications in molecular biology.

The importance of low DNA binding microcentrifuge tubes cannot be overstated. These tubes are specifically engineered to minimize the adhesion of DNA to the tube walls, which is crucial in preserving the quantity and quality of DNA samples during various laboratory procedures.

Ensuring that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results. Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (Metric) 0.6 mL
Color Natural
Material Polypropylene Resin
Volume (Metric) 0.6 mL
Sterility Non-sterile
Graduated Yes
Closure Type Snap Cap
Fit for Purpose PCR Ready
Bottom Shape Conical
Green Features ACT Ecolabel (Silver)
Shape Cylindrical
Product Type Microcentrifuge Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.