Learn More
United Scientific™ Low-Form Borosilicate Glass Beaker
SDPSCI_ED
Catalog No. S27776 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
20 L
Catalog No. Capacity (Metric)
S27776 20 L
1 options
Catalog No. S27776 Supplier United Scientific™ Supplier No. BG100020000
Add to Cart
Add to Cart
This Low-Form Borosilicate Glass Beaker features heavy and uniform wall thickness, spouts designed for easy pouring, and beaded tops.
  • Borosilicate Glass Beaker.
  • Heavy, uniform wall thickness.
  • Spout designed to minimize spillage.
  • Easy-to-read printed graduations for precise measurement.
  • Beaded Top.
Specifications
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Material Borosilicate Glass (3.3)
Type Beaker
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Scale Dual Graduated Metric Scale
Style Low Form