Learn More
*HELICA BIOSYSTEMS LOW-MATRIX AFLATOXIN
Supplier:  *HELICA BIOSYSTEMS 981AFL01LM
Encompass_Preferred
Immunochemical Assay Kits; Food Testing; Mycotoxin Detection Assay; ELISA Kit, Qualitative Low-Matrix Aflatoxin Detection, 96-strip microwell plate-based; Results in 1.5 hours, One-year stability; Excellent reproducibility
Catalog No. 50-980-590
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.