Description
- Lower gelling and melting temperature than standard agarose
- Lower gel strength than standard agarose while still easy to handle
- Higher gel transparency than standard agarose
- Greater sieving capacity than standard agarose
- Separates and finely resolves molecules >1000bp
- DNase- and RNase-Free
Specifications
Specifications
| For Use With (Application) | Used as gel matrix and preparation of large DNA samples for plugs |
| Quantity | 25 g |
| Product Type | Agarose Gel |