Learn More
IBI Scientific™ Low Melting Point Agarose
Catalog No. 15455203 Shop All Stovall Life Science Inc. Products
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-455-203 250 g
15-455-202 100 g
15-455-200 25 g
15-455-204 500 g
15-455-201 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 15-455-203 Supplier IBI Scientific™ Supplier No. IB70057
Add to Cart
Add to Cart
Designed for preparative DNA and RNA electrophoresis, cloning of tissue culture cells and viral plague assays, and fragment recovery following electrophoresis. IBI Scientific™ Low Melting Point Agarose has lower gelling point and melting point when compared to Standard Agarose allows quick and thorough digestion during extraction procedures and easy in-gel manipulations.
  • Lower gelling and melting temperature than standard agarose
  • Lower gel strength than standard agarose while still easy to handle
  • Higher gel transparency than standard agarose
  • Greater sieving capacity than standard agarose
  • Separates and finely resolves molecules >1000bp
  • DNase- and RNase-Free
Specifications
For Use With (Application) Used as gel matrix and preparation of large DNA samples for plugs
Quantity 250 g
Product Type Agarose Gel