Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Low pl Marker Kit

Catalog No. 09920059 Shop All Cytiva Products
Change view
Click to view available options
pH Range:
3.5 to 6.5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
09-920-059 3.5 to 6.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-920-059 Supplier Cytiva Supplier No. 29443548
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The markers may be run in parallel with the experimental samples on isoelectric focusing gels.

  • pl markers are lyophilized mixtures of stable, salt-free, highly purified proteins
  • These markers are not prestained
  • Amylglucosidase - 3.50 pI (Native)
  • Methyl red (dye) - 3.75 pI (Native)
  • Glucose oxidase - 4.25 pI (Native)
  • Trypsin inhibitor - 4.55 pI (Native)
  • b-Lactoglobulin A - 5.20 pI (Native)
  • Carbonic anhydrase B (bovine) - 5.85 pI (Native)
  • Carbonic anhydrase B (human) - 6.55 pI (Native)
  • Not for use in denaturing (urea) systems
  • Store at 4°C
TRUSTED_SUSTAINABILITY

Specifications

Color White
pH Range 3.5 to 6.5
For Use With (Application) To be used for isoelectric focusing (IEF)
Storage Requirements 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.