Learn More
Thermo Scientific™ Low Profile Tubes, strips of 8, Sustain™ Series
GREENER_CHOICE
Catalog No. FERAB0771SS Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
120 Strips
Catalog No. Quantity
FERAB0771SS 120 Strips
1 options
Catalog No. FERAB0771SS Supplier Thermo Scientific™ Supplier No. AB0771SS
Add to Cart
Add to Cart
Thermo Scientific™ Low-Profile Tubes, strips of 8, Sustain Series, reduce the 'dead space' between the heated lid of the thermal cycler and the sample.
Thermo Scientific™ Low-Profile Tubes, strips of 8, Sustain Series, reduce the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Low profile products are especially recommended for use with reaction volumes below 20 μL.
Features and benefits
  • Suitable for 0.1 mL thermal cycler blocks
  • 150 μL maximum tube capacity when capped
  • Low profile to reduce dead space and increase PCR efficiency
  • Labelled A–H end tabs (8-strip)
  • Sealable with flat, domed, and ultra-clear cap strips
  • Certified DNase-, RNase-, human DNA-, and PCR inhibitor-free
A breakthrough in sustainable PCR technology
Thermo Scientific™ Sustain Series PCR plastics signify a breakthrough in our PCR and quantitative PCR (qPCR) product offering that goes beyond providing accurate and reproducible PCR data. They represent our commitment to integrating more sustainable practices into our manufacturing processes, without compromising on product quality or performance. Our Sustain Series PCR plastics are manufactured under ISO 9001 guidelines within a Class 100,000 cleanroom from a biobased medical-grade virgin polypropylene that is chemically and molecularly identical to our existing Thermo Scientific™ PCR plastics. These products are supported by certification from the globally recognized International Sustainability and Carbon Certification (ISCC) system. This greener approach is equivalent to 1.96X lower CO2 equivalent emissions per kg of biobased polypropylene resin compared to traditional fossil fuel-based polypropylene resin.
Additional product details
  • Uniform, ultra-thin walls enable maximum and consistent heat transfer for equally high performance of every sample
  • Each well is visually inspected and electrostatically tested for defects and contaminants, and every lot is run through a PCR cycling test to evaluate sealing
  • Manufactured from second generation bio-circular feedstocks that are chemically and molecularly identical to our existing Thermo Scientific™ PCR plastics
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free
Color Natural
Quantity 120 Strips
Material Polypropylene
Closure Type Not Included
Autoclavable Autoclavable
Green Features ISCC PLUS
Who is the ISCC and what is ISCC PLUS?
The ISCC is an independent initiative and globally recognized certification system supporting more sustainable, traceable, and deforestation-free supply chains. It sets standards and provides certification for various sectors, including agriculture, food, feed, chemicals, and biofuels. The ISCC aims to promote more sustainable production and trade while addressing environmental and social aspects. ISCC PLUS certification is one of the certification schemes under the ISCC certification system. One focus area under this scheme is on sustainability and traceability aspects of biomass-based products, such as biofuels, bioplastics, and biochemicals. ISCC PLUS certification verifies chain of custody, traceability throughout the supply chain, and compliance with sustainability criteria through independent third-party certification. It provides assurance to customers and stakeholders that the certified products meet strict sustainability requirements.
What are ''bio-circular'' feedstocks?
Bio-circular feedstocks, a feedstock category defined by the International Sustainability and Carbon Certification (ISCC) certification system, refer to materials that are considered waste or processing residue at the beginning of the supply chain that are not landfilled or energetically used, but instead reused, further used, or recycled in a loop without dropping out of the economy. These feedstocks can be used to make biobased plastics without sacrificing performance and have a lower environmental footprint compared to virgin fossil fuel-based feedstocks.
Do plastics with biobased content allocated on a mass balance basis impact performance?
Incorporation of biobased and fossil fuel-based resins occurs at the monomer level, which helps ensure that the properties and characteristics of the biobased plastics remain consistent with traditional fossil fuel-based plastics. Plastics with biobased content by mass balance reduce reliance on fossil fuel-based resources while maintaining desired performance.
What are the benefits of purchasing biobased plastics?
Biobased plastic products from Thermo Fisher Scientific are indistinguishable from traditional fossil fuel-based plastics and have a lower carbon footprint without sacrificing performance. Additionally, there is no need for revalidation or retesting, as the product is chemically and molecularly identical to the existing fossil fuel-based version.
What are biobased plastics?
Biobased plastics are fully or partly made from biological raw materials, as opposed to the petroleum or fossil fuel used in conventional plastics.