Learn More
Azure Biosystems Low Profile White Thin-Wall 8-Tube Strip, 0.1 mL
SDP
Catalog No. AIQ323AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Color:
White
Catalog No. Color
AIQ323AZ White
1 options
Catalog No. AIQ323AZ Supplier Azure Biosystems Supplier No. AIQ323
Add to Cart
Add to Cart
Low-profile, white, opaque, 8-tube strips (0.1 mL). Pack of 120.
  • 0.1 mL tube strips are specially designed for qPCR applications and assays. Caps sold separately.
  • Thin wall
  • Low profile
  • White, opaque,
  • 0.1 mL
  • Pack of 120
Specifications
Color White
Volume (Metric) 0.1 mL
Sterility Non-sterile
Material Polypropylene
Closure Type Inverted Dome
Autoclavable Not Autoclavable
Closure Included No
Disposable Single-use