Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Low-Retention Pipet Tips, Nonfilter

Catalog No. 02717136 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
10 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
02-717-136 10 μL
02-717-137 0.1 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-717-136

Increase accuracy by reducing sample retention

  • Low retention surface minimizes samples loss
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Natural color
TRUSTED_SUSTAINABILITY

Specifications

Description Low retention Non-sterile, Bulk
Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Model Low retention
Format Bulk
Tip Style Low Retention, Extended Length
For Use With (Equipment) Eppendorf
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.