Learn More
RWD LIFE SCIENCE INC LOW STRESS ANESTH IND CHAMBER
SDP
Supplier:  RWD LIFE SCIENCE INC V105
Encompass_Preferred
Low Stress Anesthesia Induction Chamber, Mice and Rats (29cm x 15cm x 17cm), The pipeline connecting Gthe Gas Evacuation Apparatus is already include; Rodents lack cone cells that sense red light and are insensitive to red light.The red transparent appearance is convenient for the experimenter to observe the stateof the animal while reducing the stress response of the animal and reducing the impacton the biological rhythm of the animal. Comply with animal welfare.
Catalog No. NC2946611
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.