Learn More
Perkin Elmer US LLC Low Temperature Sampling Accessory
Supplier:  Perkin Elmer US LLC N4201045
Encompass_Preferred
The controllable temperature range for the Low Temperature Sampling Accessory is -196 °C RT. This accessory is compatible with the FL 6500/8500 fluorescence spectrometers.
Catalog No. 50-211-2362
Only null left
Add to Cart
Request a Quote