Learn More
Vikan™ Low Undercarriage
Catalog No. 17227656 Shop All Remco Products
Change view
Click to view available options
Color:
Steel
Catalog No. Color
17-227-656 Steel
1 options
Catalog No. 17-227-656 Supplier Vikan™ Supplier No. 6904
Only null left
Add to Cart
Add to Cart
Low Undercarriage is a solution for easily moving carts on floors that may be covered in animal grease or debris.
  • Galvanized steel low undercarriage
  • Features caster wheels that are impervious to meat fat and bearings equipped with grease fittings
  • Use a low undercarriage when mobility is required, but tipping is not necessary
  • This tub accessory meets all USDA specifications and is commonly used for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications
  • Replacement casters and wheels are available
  • The steel construction means the undercarriage will last even when exposed to the chemicals and substances commonly used in processing plants
  • Color-coded
Specifications
Product Type Undercarriage
Color Steel
Material Galvanized Steel
Quantity 1
Dimensions (L x W x H) 40.25 x 26.9 x 8.9 in.