Learn More
MilliporeSigma™ Supelco™ Low Volume Static Mixer (50 to 250 μL)
$542.50 - $666.00
Specifications
For Use With (Equipment) | ASI Low Volume Mixer Housing |
---|---|
Volume (Metric) | 50 to 250 μL |
Catalog Number | Mfr. No. | Description | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Material | Price | Quantity & Availability | ||||
11-101-6301
|
MilliporeSigma™ Supelco™
57545 |
50 μL cartridge | Stainless Steel |
Each for $548.50
|
|
||||
11-100-3809
|
MilliporeSigma™ Supelco™
57546 |
150 μL cartridge | Stainless Steel |
Each for $542.50
|
|
||||
11-100-3709
|
MilliporeSigma™ Supelco™
500461 |
250 μL cartridge | PEEK |
Each for $666.00
|
|
||||
11-100-3803
|
MilliporeSigma™ Supelco™
57547 |
250 μL cartridge | Stainless Steel |
Each for $546.00
|
|
||||
Description
Specifications
ASI Low Volume Mixer Housing | |
50 to 250 μL |