Learn More
Radians Lowset™ 21 Earmuffs
SDP
Radians Lowset™ 21 Earmuffs have foam filled, cushion padding on earcups along with evenly spread tension in the headband to allow for comfortable wear while wearing protective safety glasses
$22.95
Specifications
Cushion Material Foam
Hearing Protection Style Passive Ear Muff
For Use With (Application) Job Site Applications Including Shooting, Hunting, Construction, Manufacturing, Oil and Gas, Warehousing, Lawn and Garden, Automotive, Assembly, Public Safety, Home and DIY, or Utilities
Model Lowset™
Quantity 1 Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Cushion Color Price Quantity & Availability  
Catalog Number Mfr. No. Cushion Color Price Quantity & Availability  
19-003-084
SDP
View Documents
Radians
LS0800CS
Pink
Each for $22.95
Case of 24 EA for $456.50
Only null left
Add to Cart
19-003-083
SDP
View Documents
Radians
LS0100CS
Black N/A N/A N/A
Description
Description
  • Comfortable and lightweight hearing protection
  • Low profile design
  • Compact folding feature allows for easy storage and keeps particles out of the earcups
  • Adjustable headband with side wire guides allows for optimum fit
  • Padded moisture wicking headband moves moisture away while providing cool comfort
Specifications
Specifications
Foam
Job Site Applications Including Shooting, Hunting, Construction, Manufacturing, Oil and Gas, Warehousing, Lawn and Garden, Automotive, Assembly, Public Safety, Home and DIY, or Utilities
1 Pk.
No
No
No
Passive Ear Muff
Lowset™
21 dB
Reusable
Black
Yes
Videos
SDS
Documents
Documents
Product Certifications
WARNING: Cancer and reproductive harm - www.P65warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.