Learn More
Invitrogen™ LOX Polyclonal Antibody

Description
This is a highly glycosylated protein and can exhibit bands at ~32 kDa, ~50 kDa and ~58 kDa representing the mature (secreted) form, the pro form and the glycosylated forms, respectively. Suggested positive control: MC3T3-E1 lysate.
Specifications
Specifications
| Antigen | LOX |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | LOX |
| Gene Accession No. | P16636, P28300, P28301, P45845 |
| Gene Alias | AI893619; cross-linking enzyme; H-rev142; LOX; Lysyl oxidase; Lysyl oxidase (an H-rev gene with its expression down-regulated in HRAS-transformed rat 208F fibroblasts); MGC105112; protein-lysine 6-oxidase; Protein-lysine 6-oxidase, long form; Protein-lysine 6-oxidase, short form; Ras excision protein; ras recision; rrg; Rrg1; TSC-160 |
| Gene Symbols | LOX |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.