Learn More
AdipoGen Loxoribine, 121288-39-9, MFCD00867591, 5 mg
SDP
Supplier:  AdipoGen AGCR13584M005
Encompass_Preferred
MF: C13H17N5O6, MW: 339.3, ≥97%, Appearance: White to off-white powder. Guanosine analog derivatized at position N7 and C8. Very potent immunostimulatory compound that increases cytokine production. Antitumor and antiviral activity compound. Potent and selective synthetic ligand for Toll-like receptor 7 (TLR7). Co-incubation with thymidine homopolymer oligodeoxynucleotides (ODN) redirected the stimulatory effect of loxoribine away from TLR7 toward TLR8. Induces autophagy and autophagy-induced cell death in Caco-2 cells. Shows adjuvant activity to increase the effectiveness of vaccines.
Catalog No. 50-117-6103
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.