Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LP 20 hydrochloride, Tocris Bioscience™

Catalog No. 470450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-045-0 50 mg
47-041-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-045-0 Supplier R&D Systems Supplier No. 4704/50
This item is not returnable. View return policy

Selective 5-HT7 ligand

Ligand of the 5-HT7 receptor (Ki = 2.6 nM); exhibits agonist and antagonist activity. Displays selectivity over adrenergic α1 and 5-HT1A receptors (Ki values are 156 and 476 nM respectively).

Chemical Identifiers

CAS 1386928-34-2
Molecular Formula C17H21ClN2O
Molecular Weight (g/mol) 304.818
InChI Key KXZUJPBDJNUNCN-UHFFFAOYSA-N
Synonym lp 20 hydrochloride, lp-20 hydrochloride, unii-2646g1d2wa, 1-4'-methoxy 1,1-biphenyl-2-yl piperazine hydrochloride, 1-4'-methoxy 1,1'-biphenyl-2-yl-piperazine hydrochloride, piperazine, 1-4'-methoxy 1,1'-biphenyl-2-yl-, hydrochloride 1:1, 1-4'-methoxy-1,1'-biphenyl-2-yl piperazine hydrochloride
PubChem CID 22001617
IUPAC Name 1-[2-(4-methoxyphenyl)phenyl]piperazine;hydrochloride
SMILES COC1=CC=C(C=C1)C2=CC=CC=C2N3CCNCC3.Cl

Specifications

Quantity 50 mg
Formula Weight 304.81
Percent Purity >98%
Chemical Name or Material LP 20 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.