Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Broadpharm LP01, 25 mg, Ionizable lipid, siRNA, mRNA, DNA, biodegradable, -20°C storage, ambient shipping, 95% purity

Supplier:  Broadpharm BP2680925MG

Encompass

LP01 is a biodegradable ionizable lipid designed for efficient and safe nucleic acid and small molecule delivery via lipid nanoparticles (LNPs). Its labile ester linkages significantly reduce liver bioaccumulation compared to conventional ionizable lipids, making it well-tolerated for in vivo applications. LP01 facilitates the encapsulation and intracellular delivery of various nucleic acids and small molecules, enabling targeted therapeutic applications with enhanced safety profiles.

  • Biodegradable via labile ester linkages
  • Reduces liver bioaccumulation
  • Encapsulates siRNA, mRNA, DNA, and small molecules
  • Storage condition at -20°C
  • Shipping at ambient temperature
  • Availability is in stock
  • Ships within 24 hours
  • Soluble in ethanol, DMSO, DMF
  • Standard quantity of 25 mg

Catalog No. NC2682691


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.