Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPCAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596301 Supplier Invitrogen™ Supplier No. PA596301
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPSMVSRNEN AQVPLIGRLL RAVQPVLVSR VDPDSRKNTI NEIIKRTTSG GEWPQILVFP EGTCTNRSCL ITFKPGAFIP GVPVQPVLLR YPNKLDTVTW TWQGYTFIQL CMLTFCQLFT KVEVEFMPVQ VPNDEEKNDP VLFANKVRNL MAEALGIPVT DHTYEDCRLM ISAGQLTLPM EAGLVEFTKI SRKLKLDWDG VRKHLDEYAS IASSSKGGRI GIEEFAKYLK LPVSDVLRQL FALFDRNHDG SIDFREYVIG LAVLCNPSNT EEIIQVAFKL FDVDEDGYIT EEEFSTILQA SLGVPDLDVS GLFKEIAQGD SISYEEFKSF ALKHPEYAKI FTTYLDLQTC HVFSLPKEVQ TTPSTASNKV SPEKHEESTS DKKDD; Positive Samples: HeLa, Jurkat; Cellular Location: Endoplasmic reticulum membrane, Golgi apparatus membrane, Lipid droplet, Single-pass type II membrane protein.

This gene encodes a member of the lysophospholipid acyltransferase family. The encoded enzyme may function in two ways: to catalyze the biosynthesis of platelet-activating factor (1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine) from 1-O-alkyl-sn-glycero-3-phosphocholine, and to catalyze the synthesis of glycerophospholipid precursors from arachidonyl-CoA and lysophosphatidylcholine. The encoded protein may function in membrane biogenesis and production of platelet-activating factor in inflammatory cells. The enzyme may localize to the endoplasmic reticulum and the Golgi. [provided by RefSeq, Feb 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPCAT2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LPCAT2
Gene Accession No. Q7L5N7
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 11; 1-acylglycerophosphocholine O-acyltransferase; 1-AGP acyltransferase 11; 1-AGPAT 11; 1-alkylglycerophosphocholine O-acetyltransferase; A330042H22; Acetyl-CoA:lyso-PAF acetyltransferase; acetyl-CoA:lyso-platelet-activating factor acetyltransferase; acyl-CoA:lysophosphatidylcholine acyltransferase 2; acyl-CoA:lysophosphatidylcholine acyltransferase member 1; acyltransferase like 1; acyltransferase like 1A; acyltransferase-like 1; acyltransferase-like 1-A; AGPAT11; AYTL1; Aytl1a; LPAAT-alpha; lpafat1; LPC acyltransferase 2; Lpcat2; LPCAT-2; Lpcat2a; lyso-PAF acetyltransferase; LysoPAFAT; lysoPAFAT/LPCAT2; lysoPC acyltransferase 2; lysophosphatidic acid acyltransferase alpha; Lysophosphatidylcholine acyltransferase 2; lyso-platelet-activating factor (PAF) acetyltransferase; RGD1563994
Gene Symbols LPCAT2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-544 of human LPCAT2 (NP_0603092).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54947
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.