Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPCAT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. p-200128787
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119263 200 μL
PIPA5121793 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119263 Supplier Invitrogen™ Supplier No. PA5119263
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LPCAT3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

LPCAT3 (Lysophosphatidylcholine Acyltransferase 3) is a Protein Coding gene. Diseases associated with LPCAT3 include Retinitis Pigmentosa 40 and Fanconi Anemia, Complementation Group D2. Among its related pathways are triacylglycerol biosynthesis and Glycerophospholipid biosynthesis. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring acyl groups and 1-acylglycerophosphocholine O-acyltransferase activity. An important paralog of this gene is MBOAT1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPCAT3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene LPCAT3
Gene Accession No. Q6P1A2
Gene Alias Lysophosphatidylcholine Acyltransferase 3
Gene Symbols LPCAT3
Host Species Rabbit
Immunogen Recombinant protein Lysophosphatidylcholine Acyltransferase 3. The antigen corresponds to amino acid range 122-233 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10162
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.