Learn More
MilliporeSigma™ Supelco™ LpDNPH Ozone Scrubber S10L (Reversible Cartridge), Potassium Iodide (KI)
Catalog No. 111010864 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
11-101-0864 10/Pk.
1 options
Catalog No. 11-101-0864 Supplier MilliporeSigma™ Supelco™ Supplier No. 505285
Add to Cart
Add to Cart
Ozone scrubbers are recommended for sampling carbonyls in high humidity environments.
LpDNPH cartridges contain 1.5 g of high purity potassium iodide (KI) and are available in two cartridge styles. KI traps the ozone which causes a negative formaldehyde interference in DNPH coated devices. The reversible cartridge offers a slip luer end-fitting, and the rezorian cartridge offers a luer lock end-fitting, to enable you to connect your cartridge of choice directly to the inlet of any DNPH cartridge with a like end-fitting. Testing (200 ppb ozone, 50 % RH, 25C) indicates the ozone scrubber to have an ozone capacity of 100,000 ppb/hr.
TRUSTED_SUSTAINABILITY
Specifications
Description Potassium Iodide (KI), 1.5 g
Quantity 10/Pk.