Learn More
MilliporeSigma™ Supelco™ LpDNPH S10 Cartridge, Bed wt. 350 mg
Catalog No. 111010101 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Type:
S10 Cartridge
SPE Cartridge
Quantity:
10/Pk.
50/Pk.
Packaging:
1/Zip Bag
Bulk Packed
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity Packaging
11-101-0101 SPE Cartridge 10/Pk. Bulk Packed
11-100-8039 SPE Cartridge 50/Pk. Bulk Packed
11-101-6363 S10 Cartridge 50/Pk. Bulk Packed
11-100-8181 SPE Cartridge 50/Pk. 1/Zip Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-101-0101 Supplier MilliporeSigma™ Supelco™ Supplier No. 21026U
Add to Cart
Add to Cart
Easy to use in the field and laboratory
The design of the S10 cartridge makes it easy to use in the field and laboratory. Reusable adaptors are available for connecting the cartridge to the sampling pump. The built in reservoir eliminates the need to attach a syringe for sample extraction/elution.
TRUSTED_SUSTAINABILITY
Specifications
Basis Weight 350 mg
Certifications/Compliance ASTM D5197/EPA 100,IP-6 A,TO-11 A/NIOSH 2016
Packaging Bulk Packed
Quantity 10/Pk.
Type SPE Cartridge
Volume (Metric) 3 mL