Learn More
Invitrogen™ LPL Polyclonal Antibody

Description
This antibody detects human Lipoprotein Lipase/LPL in direct ELISAs and less than 1% cross-reactivity with recombinant human (rh) LIPG, rhLIPI, and rhPNLIPRP1 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
Specifications
Specifications
| Antigen | LPL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | LPL |
| Gene Accession No. | P06858, P11152 |
| Gene Alias | adipose lipoprotein lipase; alpha lipase; HDLCQ11; I79_006077; LIPD; lipoprotein lipase; lipoprotein lipase (EC 3.1.1.34); lipoprotein lipase precursor; lipoprotein lipase preprotein; Lipoprotein lipase-like protein; LPL; O 1-4-5; phospholipase A1; triacylglycerol lipase |
| Gene Symbols | LPL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.