Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lplunc1 Monoclonal Antibody (4H2)
GREENER_CHOICE

Catalog No. PIMA517124
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517124 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517124 Supplier Invitrogen™ Supplier No. MA517124
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17124 targets Lplunc1 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human samples. The MA5-17124 immunogen is purified recombinant fragment of mouse Lplunc1 expressed in E. Coli. MA5-17124 detects Lplunc1 which has a predicted molecular weight of approximately 52.4kDa.

The protein encoded by this gene may be involved in the innate immune response to bacterial exposure in the mouth, nasal cavities, and lungs. The encoded protein is secreted and is a member of the BPI/LBP/PLUNC protein superfamily. This gene is found with other members of the superfamily in a cluster on chromosome 20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lplunc1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4H2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene BPIFB1
Gene Accession No. Q61114, Q8TDL5
Gene Alias BPI fold containing family B member 1; BPI fold containing family B, member 1; BPI fold-containing family B member 1; BPIFB1; C20orf114; long palate, lung and nasal epithelium carcinoma associated 1; long palate, lung and nasal epithelium carcinoma-associated protein 1; Lplunc1; UNQ706/PRO1357; VEMSGP; von Ebner minor salivary gland protein; von Ebner minor salivary protein
Gene Symbols BPIFB1
Host Species Mouse
Immunogen Purified recombinant fragment of mouse Lplunc1 expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228801, 92747
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.