Learn More
Invitrogen™ Lplunc1 Polyclonal Antibody

Description
Immunogen sequence: PKVIKEKLTQ ELKDHNATSI LQQLPLLSAM REKPAGGIPV LGSLVNTVLK HIIWLKVITA NILQLQVKPS ANDQELLVKI PLDMVAGFNT PLVKTIVEFH MTTEAQATIR MDTSASGPTR LVLSDCATSH Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 64%.
Specifications
Specifications
| Antigen | Lplunc1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BPIFB1 |
| Gene Accession No. | Q8TDL5 |
| Gene Alias | BPI fold containing family B member 1; BPI fold containing family B, member 1; BPI fold-containing family B member 1; BPIFB1; C20orf114; long palate, lung and nasal epithelium carcinoma associated 1; long palate, lung and nasal epithelium carcinoma-associated protein 1; Lplunc1; UNQ706/PRO1357; VEMSGP; von Ebner minor salivary gland protein; von Ebner minor salivary protein |
| Gene Symbols | BPIFB1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.