Learn More
Invitrogen™ LPS Monoclonal Antibody (C6)
Description
LPS Monoclonal Antibody for IHC (P), Western Blot
Specifications
Specifications
| Antigen | LPS |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | C6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene Alias | LPS |
| Host Species | Mouse |
| Immunogen | OVA Conjugated Lipopolysaccharide (LPS) from Escherichia coli (O55:B5). |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.