Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531046
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531046 Supplier Invitrogen™ Supplier No. PA531046
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: human plasma. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The leucine-rich repeat (LRR) family of proteins, including LRG1, have been shown to be involved in protein-protein interaction, signal transduction, and cell adhesion and development. LRG1 is expressed during granulocyte differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRG1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene LRG1
Gene Accession No. P02750
Gene Alias 1300008B03Rik; 2310031E04Rik; A2GL; HMFT1766; leucine rich alpha 2 glycoprotein; leucine rich alpha-2-glycoprotein 1; leucine-rich alpha-2-glycoprotein; leucine-rich alpha-2-glycoprotein 1; Leucine-rich alpha-2-glycoprotein precursor; LRG; LRG 1; LRG1; LRG-1; Lrhg
Gene Symbols LRG1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human LRG1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116844
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.