Learn More
Tocris Bioscience™ LRGILS-NH2
Catalog No. 33941
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
33-941 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 33-941 Supplier Tocris Bioscience™ Supplier No. 3394/1
This item is not returnable. View return policy
Control peptide for SLIGRL-NH2 (Cat. No. 1468)
LRGILS-NH2 is a reversed amino acid sequence control peptide for SLIGRL-NH2, a protease-activated receptor-2 (PAR2) agonist that facilitates gastrointestinal transit in vivo.
Active Analog also available.
Specifications
Quantity 1 mg