Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553323
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553323 Supplier Invitrogen™ Supplier No. PA553323
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CPKGCLCSSS GGLNVTCSNA NLKEIPRDLP PETVLLYLDS NQITSIPNEI FKDLHQLRVL NLSKNGIEFI DEHAFKGVAE TLQTLDLSDN RIQSVHKNAF NNLKARARIA NNPWHCDCTL QQVLRSMASN HETAHNVICK TSVLD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The protein encoded by this gene is a tumor suppressor, with lowered expression levels found in gastric, renal, colorectal, lung, and breast cancer tissues. The promoter of this gene is frequently hypermethylated in these cancer tissues, although the hypermethylation does not appear to be the cause of the reduced expression of this gene. Several transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC3B
Gene Accession No. Q96PB8
Gene Alias BC019794; leucine rich repeat containing 3B; leucine-rich repeat protein LRP15; Leucine-rich repeat-containing protein 3B; LRP15; LRRC3B; RGD1311897; UNQ195/PRO221
Gene Symbols LRRC3B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRP15. Recombinant protein control fragment (Product #RP-90526).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116135
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.